missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, 99%

Product Code. 10307360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307360 25 mL Glass bottle 25mL
10449650 100 mL Glass bottle 100mL
13438418 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307360 Supplier Thermo Scientific Acros Supplier No. 176030250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-86-5 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00003250
InChI Key GSNUFIFRDBKVIE-UHFFFAOYSA-N
Synonym furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
PubChem CID 12266
IUPAC Name 2,5-dimethylfuran
SMILES CC1=CC=C(O1)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless to Amber
Density 0.9000g/mL
Boiling Point 92.0°C to 94.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 25 mL
Beilstein 17,41
Merck Index 15,3268
Specific Gravity 0.9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in oil,organic solvents
Formula Weight 96.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylfuran, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethylfuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.