missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzylamine, 98%, Thermo Scientific Chemicals

Product Code. 10773973
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-48-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00039780
InChI Key LUJNPFWZXIGIPS-UHFFFAOYSA-N
PubChem CID 66735
IUPAC Name (2,5-dimethylphenyl)methanamine
SMILES CC1=CC(=C(C=C1)C)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.538
Quantity 1 g
Synonym 2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
Formula Weight 135.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylbenzylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'