missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzyl chloride, 98%

Product Code. p-7043560
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11364544

Brand: Thermo Scientific Alfa Aesar L01303.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-45-3 | C9H11Cl | 154.637 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-45-3
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00000902
InChI Key PECXPZGFZFGDRD-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzyl chloride, 2-chloromethyl-1,4-dimethylbenzene, 1-chloromethyl-2,5-dimethylbenzene, 2-chloromethyl-p-xylene, benzene, 2-chloromethyl-1,4-dimethyl, 2-chloromethyl-1,4-dimethyl-benzene, benzenemethanol,2-bromo-5-fluoro, 2,5-dimethylbenzylchloride, acmc-209poz, 2.5-dimethylbenzylchloride
PubChem CID 69987
IUPAC Name 2-(chloromethyl)-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)CCl

Specifications

Density 1.05
Boiling Point 232°C to 233°C
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.537
Quantity 5 g
UN Number UN3265
Beilstein 2206277
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylbenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.