missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzoic acid, 98%

Product Code. 10542181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10542181 10 g Glass bottle 10g
10187253 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10542181 Supplier Thermo Scientific Acros Supplier No. 160330100
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 610-72-0 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-72-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002482
InChI Key XZRHNAFEYMSXRG-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dimethyl, isoxylic acid, 2,5-dimethyl benzoic acid, unii-rke5pmk0h6, 2,5-dimethyl-benzoic acid, 2-carboxy-1,4-dimethylbenzene, rke5pmk0h6, p-xylene-2-carboxylic acid, 2,5-dimethylbenzenecarboxylic acid, 2,5-dimethylbenzoicacid
PubChem CID 11892
ChEBI CHEBI:64825
IUPAC Name 2,5-dimethylbenzoic acid
SMILES CC1=CC(=C(C=C1)C)C(=O)O

Specifications

Melting Point 131.0°C to 135.0°C
CAS Min % 97.5
Color White
CAS Max % 100.0
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 534
Boiling Point 268.0°C
Solubility Information Solubility in water: insoluble
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2, 5-Dimethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.