missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10262882
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10262882 10 g Amber Glass Bottle 10g
10672253 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262882 Supplier Thermo Scientific Maybridge Supplier No. MO07001EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19040-62-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00024875
InChI Key FZVZUIBYLZZOEW-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
PubChem CID 87910
IUPAC Name 2,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1)C)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Quantity 10 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula (CH3)2C6H3SO2Cl
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylbenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.