missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-thiaadipic Acid, TRC

Product Code. 30369532
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369532 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369532 Supplier Toronto Research Chemicals Supplier No. TRCD469600100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 106014-16-8
Molecular Formula C7H12O4S
Quantity 100 mg
Synonym 3-[(1-Carboxyethyl)thio]-2-methyl-propanoic Acid
InChI Formula InChI=1S/C7H12O4S/c1-4(6(8)9)3-12-5(2)7(10)11/h4-5H,3H2,1-2H3,(H,8,9)(H,10,11)
SMILES OC(C(C)SCC(C)C(O)=O)=O
IUPAC Name 3-((1-carboxyethyl)thio)-2-methylpropanoic acid
Molecular Weight (g/mol) 192.23
Formula Weight 192.05
Chemical Name or Material 2,5-Dimethyl-3-thiaadipic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.