missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™

Product Code. 10337250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337250 5 g Glass bottle 5g
10104470 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10337250 Supplier Thermo Scientific Acros Supplier No. 165930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59480-92-1
Molecular Formula C6H12N
Molecular Weight (g/mol) 98.17
MDL Number MFCD00005215
InChI Key TXQDHQBSNAJSHQ-OLQVQODUSA-O
PubChem CID 101066
IUPAC Name 2,5-dimethyl-2,5-dihydro-1H-pyrrole
SMILES C[C@H]1[NH2+][C@@H](C)C=C1

Specifications

CAS Min % 70.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8240g/mL
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Refractive Index 1.4404
Quantity 5 g
Specific Gravity 0.824
Synonym 2,5-dimethyl-3-pyrroline, 2,5-dimethyl-3-pyrroline, mixture of cis and trans, 2,5-dimethyl-2,5-dihydropyrrole, 1h-pyrrole,2,5-dihydro-2,5-dimethyl, 1h-pyrrole, 2,5-dihydro-2,5-dimethyl
Formula Weight 97.16
Percent Purity 75%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethyl-3-pyrroline, 75%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethyl-3-pyrroline, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.