missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 70.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 0.8240g/mL |
Flash Point | 5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 70% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4404 |
Quantity | 1 g |
Show More |
Safety and Handling
Product Identifier
- 2,5-Dimethyl-3-pyrroline, mixture of cis and trans
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'