missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-hexyne-2,5-diol, 98%, Thermo Scientific™

Product Code. 10696422
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 142-30-3
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
MDL Number MFCD00004468
InChI Key IHJUECRFYCQBMW-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
PubChem CID 8883
IUPAC Name 2,5-dimethylhex-3-yne-2,5-diol
SMILES CC(C)(C#CC(C)(C)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >89.0°C
Color White to Yellow
Boiling Point 121.0°C to 123.0°C (7.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC(CH3)2C≡CC(CH3)2OH
Quantity 100 g
Beilstein 01, 501
Solubility Information Solubility in water: 240g/L (20 c). Other solubilities: soluble in acetone,alcohol and ethyl acetate,slightly soluble in benzene,naphtha and,carbon tetrachloride
Formula Weight 142.2
Percent Purity 98%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2, 5-Dimethyl-3-hexyne-2, 5-diol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'