missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-furansulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10150872
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10150872 250 mg Amber Glass Bottle 250mg
10501692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10150872 Supplier Thermo Scientific Maybridge Supplier No. CC01103CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-26-7
Molecular Formula C6H7ClO3S
Molecular Weight (g/mol) 194.63
MDL Number MFCD04115370
InChI Key PIXAQZAVANAGLE-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-furansulfonyl chloride, 2,5-dimethyl-furan-3-sulfonylchloride, 2,5-dimethylfuran 3-sulfonyl chloride, 2,5-dimethyl 3-furyl chlorosulfone, 3-furansulfonylchloride, 2,5-dimethyl
PubChem CID 2795024
IUPAC Name 2,5-dimethylfuran-3-sulfonyl chloride
SMILES CC1=CC(=C(C)O1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.64
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-3-furansulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.