missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3(2H)-furanone, 94%

Product Code. 11474823
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474823 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474823 Supplier Thermo Scientific Alfa Aesar Supplier No. A12182.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14400-67-0 | C6H8O2 | 112.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14400-67-0
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00052571
InChI Key ASOSVCXGWPDUGN-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3 2h-furanone, 2,5-dimethylfuran-3 2h-one, 2,5-dimethyl-2,3-dihydrofuran-3-one, 3 2h-furanone, 2,5-dimethyl, 2,5-dimethyl-3 2h furanone, 2,5-dimethyl-2h-furan-3-one, 2,5-dimethyl-3-2h-furanone, 2h-furan-3-one, 2,5-dimethyl, mango furanone
PubChem CID 85730
IUPAC Name 2,5-dimethylfuran-3-one
SMILES CC1C(=O)C=C(O1)C

Specifications

Density 1.06
Boiling Point 259°C to 261°C
Flash Point 68°C (154°F)
Odor Sweet
Quantity 25 g
Formula Weight 112.13
Percent Purity 94%
Chemical Name or Material 2,5-Dimethyl-3 (2H)-furanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.