missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-hexanediol, 97%

Product Code. 11434024
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434024 2500 g 2500g
11414024 100 g 100g
11424024 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434024 Supplier Thermo Scientific Alfa Aesar Supplier No. A11984.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-03-2 | C8H18O2 | 146.23 g/mol

It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

Solubility
Soluble in water 20°C 140g/L.

Notes
Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-03-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004473
InChI Key ZWNMRZQYWRLGMM-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
PubChem CID 8031
IUPAC Name 2,5-dimethylhexane-2,5-diol
SMILES CC(C)(O)CCC(C)(C)O

Specifications

Melting Point 87°C to 91°C
Density 0.898
Boiling Point 214°C to 215°C
Flash Point 126°C (258°F)
Odor Odorless
Quantity 2500 g
Beilstein 1361437
Solubility Information Soluble in water 20°C 140g/L.
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 2,5-Dimethyl-2,5-hexanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.