Learn More
2,5-Dimethyl-2,5-hexanediol, 97%
Description
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.
Solubility
Soluble in water 20°C 140g/L.
Notes
Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.
Specifications
Specifications
| Melting Point | 87°C to 91°C |
| Density | 0.898 |
| Boiling Point | 214°C to 215°C |
| Flash Point | 126°C (258°F) |
| Odor | Odorless |
| Quantity | 500 g |
| Beilstein | 1361437 |
| Solubility Information | Soluble in water 20°C 140g/L. |
| Formula Weight | 146.23 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
- 2,5-Dimethyl-2,5-hexanediol
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 2,5-Dimethyl-2,5-hexanediol
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.