missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,3-dihydro-1H-indole, TRC

Product Code. 30376522
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376522 50 mg -
30386116 10 mg 10mg
30376707 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376522 Supplier Toronto Research Chemicals Supplier No. TRCD89081050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90874-56-9
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
IUPAC Name 2,5-dimethyl-2,3-dihydro-1H-indole
SMILES CC1Cc2cc(C)ccc2N1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N/c1-7-3-4-10-9(5-7)6-8(2)11-10/h3-5,8,11H,6H2,1-2H3
Formula Weight 147.1048
Chemical Name or Material 2,5-Dimethyl-2,3-dihydro-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.