missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1-(pyridin-4-ylmethyl)-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10260305
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10260305 5 g 5g
10691503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10260305 Supplier Thermo Scientific Maybridge Supplier No. SP00292DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-15-2
Molecular Formula C13H14N2O2
Molecular Weight (g/mol) 230.267
MDL Number MFCD01567218
InChI Key IHQUENCAKSKBOG-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1-pyridin-4-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-pyridin-4-ylmethyl pyrrole-3-carboxylic acid, maybridge1_008545, 2,5-dimethyl-1-4-pyridylmethyl pyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-4-pyridinylmethyl, 2,5-dimethyl-1-pyridin-4-yl methyl-1h-pyrrole-3-carboxylic acid
PubChem CID 2736289
IUPAC Name 2,5-dimethyl-1-(pyridin-4-ylmethyl)pyrrole-3-carboxylic acid
SMILES CC1=CC(=C(N1CC2=CC=NC=C2)C)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 230.27
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1-(pyridin-4-ylmethyl)-1H-pyrrole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.