missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10672695
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10672695 5 g 5g
10693593 250 mg 250mg
10334432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10672695 Supplier Thermo Scientific Maybridge Supplier No. CC26801DE
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23000-14-8
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD03011595
InChI Key LHGRUGVXZLHYKE-UHFFFAOYSA-N
Synonym 2,5-dimethyloxazole-4-carboxylic acid, dimethyl-1,3-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2,5-dimethyl, 2,5-dimethyl-1,3-oxazole-4-carboxylicacid, 4-carboxy-2,5-dimethyl-1,3-oxazole, 2,5-dimethyl-oxazole-4-carboxylic acid, 2.5-dimethyl-1,3-oxazole-4-carboxylic acid
PubChem CID 2795465
IUPAC Name 2,5-dimethyl-1,3-oxazole-4-carboxylic acid
SMILES CC1=C(N=C(O1)C)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.