missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carbonyl chloride, 97%, Thermo Scientific™

Product Code. p-2697182
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Product Code. Quantity Packaging unitSize
10572852 250 mg Amber Glass Bottle 250mg
10712863 1 g Amber Glass Bottle 1g
11396825 5 g Amber Glass Bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10572852

Brand: Thermo Scientific Maybridge CC26802CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 197719-27-0
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.57
MDL Number MFCD04972642
InChI Key XZXCVPKMCPXVFF-UHFFFAOYSA-N
PubChem CID 7018020
SMILES CC1=NC(C(Cl)=O)=C(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
IUPAC Name 2,5-dimethyl-1,3-oxazole-4-carbonyl chloride
Formula Weight 159.57
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.