missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-1,3-oxazole-4-carbonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride |
IUPAC Name | dimethyl-1,3-oxazole-4-carbonyl chloride |
Formula Weight | 159.57 |
Percent Purity | 97% |
Chemical Name or Material | 2, 5-Dimethyl-1, 3-oxazole-4-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'