missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytetrahydrofuran, TRC

Product Code. 30388635
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388635 2.5 g
30377024 500 mg
30371133 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388635 Supplier Toronto Research Chemicals Supplier No. TRCD4608232.5G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(OC)O1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H12O3/c1-7-5-3-4-6(8-2)9-5/h5-6H,3-4H2,1-2H3
Formula Weight 132.0786
Chemical Name or Material 2,5-Dimethoxytetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.