missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytetrahydrofuran, cis + trans, 98%

Product Code. 11489183
Change view
Click to view available options
Quantity:
100 g
500 g
Conditionnement:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11489183 100 g 100g
11499183 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11489183 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A11687.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 696-59-3 | C6H12O3 | 132.159 g/mol

2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00005359
InChI Key GFISDBXSWQMOND-UHFFFAOYSA-N
Synonym 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf
PubChem CID 79098
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(O1)OC

Spécification

Melting Point -45°C
Density 1.02
Quantity 100 g
Refractive Index 1.418
UN Number UN3271
Beilstein 104261
Boiling Point 145°C to 147°C
Solubility Information Miscible with methanol,diethyl ether and tetrahydrofuran. Immiscible with water.
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxytetrahydrofuran, cis + trans
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.