missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytetrahydrofuran, cis + trans, 98%

Product Code. 11499183
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499183 500 g 500g
11489183 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499183 Supplier Thermo Scientific Alfa Aesar Supplier No. A11687.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 696-59-3 | C6H12O3 | 132.159 g/mol

2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00005359
InChI Key GFISDBXSWQMOND-UHFFFAOYSA-N
Synonym 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf
PubChem CID 79098
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(O1)OC

Specifications

Melting Point -45°C
Density 1.02
Quantity 500 g
Refractive Index 1.418
UN Number UN3271
Beilstein 104261
Boiling Point 145°C to 147°C
Solubility Information Miscible with methanol,diethyl ether and tetrahydrofuran. Immiscible with water.
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxytetrahydrofuran, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.