missing translation for 'onlineSavingsMsg'
Learn More

2,5-dimethoxyphenyl isothiocyanate, Thermo Scientific™

Product Code. 10352325
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10352325 5 g 5g
10037592 10 g 10g
10623275 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10352325 Supplier Thermo Scientific Maybridge Supplier No. SB01666DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40532-06-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00041059
InChI Key RADQLZMDTYWZKA-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
PubChem CID 142466
IUPAC Name 2-isothiocyanato-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(=C1)N=C=S

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 195.24
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethoxyphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.