missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11452928
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452928 50 g 50g
11448577 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452928 Supplier Thermo Scientific Alfa Aesar Supplier No. A18630.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40532-06-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00041059
InChI Key RADQLZMDTYWZKA-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
PubChem CID 142466
IUPAC Name 2-isothiocyanato-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(=C1)N=C=S

Specifications

Melting Point 35°C to 37°C
Boiling Point 174°C to 175°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN2923
Beilstein 3267310
Sensitivity Moisture sensitive
Formula Weight 195.24
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxyphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.