missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number 11402578
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
5312-97-0
C9H9NO2
163.176
MFCD00016375
HWAMEJIMXIXLIH-UHFFFAOYSA-N
79200
2,5-dimethoxybenzonitrile
COC1=CC(=C(C=C1)OC)C#N
This item is not returnable. View return policy
5312-97-0
C9H9NO2
163.176
MFCD00016375
HWAMEJIMXIXLIH-UHFFFAOYSA-N
79200
2,5-dimethoxybenzonitrile
COC1=CC(=C(C=C1)OC)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C to 84°C
Quantity 5 g
UN Number UN3439
Beilstein 2691937
Synonym benzonitrile, 2,5-dimethoxy, 2,5-dimethoxybenzenecarbonitrile, pubchem13109, acmc-1an8e, benzonitrile,2,5-dimethoxy, 2-cyano-1,4-dimethoxybenzene, 2,5-dimethoxybenzonitrile, labotest-bb lt00848082, 2-cyanohydroquinone dimethyl ether
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'