missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11413664
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1483-28-9
Molecular Formula C8H9ClO4S
Molecular Weight (g/mol) 236.67
MDL Number MFCD00024872
InChI Key SHELADVIRCCTFN-UHFFFAOYSA-N
Synonym 2,5-dimethoxybenzene-1-sulfonyl chloride, 2,5-dimethoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-dimethoxy, 2,5-dimethoxyphenyl chlorosulfone, 2,5-dimethoxybenzenesulphonylchloride, 2,5-dimethoxy-benzenesulfonyl chloride, 2,5-dimethoxybenzenesulfonylchloride, asischem d48918, pubchem5126, timtec-bb sbb003243
PubChem CID 137025
IUPAC Name 2,5-dimethoxybenzenesulfonyl chloride
SMILES COC1=CC=C(OC)C(=C1)S(Cl)(=O)=O

Specifications

Melting Point 111°C to 116°C
Quantity 5 g
UN Number UN3261
Beilstein 2648547
Sensitivity Moisture sensitive
Formula Weight 236.68
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxybenzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'