missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzaldehyde, 98+%

Product Code. 11498767
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498767 25 g 25g
11408777 100 g 100g
11418777 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498767 Supplier Thermo Scientific Alfa Aesar Supplier No. A19928.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-02-7 | C9H10O3 | 166.176 g/mol

2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-02-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003314
InChI Key AFUKNJHPZAVHGQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde
PubChem CID 66726
IUPAC Name 2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)OC)C=O

Specifications

Melting Point 47°C to 51°C
Boiling Point 146°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 509301
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.