missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxy-N-[(2-methoxyphenyl)methyl]-4-methylbenzeneethanamine Hydrochloride, TRC

Product Code. 30384362
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384362 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384362 Supplier Toronto Research Chemicals Supplier No. TRCD46222510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 25D-NBOMe (hydrochloride)
CAS 1539266-35-7
Molecular Formula C19 H25 N O3 . Cl H
Quantity 10 mg
Synonym 25D-NBOMe HCl, 2-(4-Methyl-2,5-dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine Hydrochloride, 2-(2,5-Dimethoxy-4-methyl-phenyl)-N-[(2-methoxyphenyl)methyl]ethanamine hydrochloride, Benzeneethanamine, 2,5-dimethoxy-N-[(2-methoxyphenyl)methyl]-4-methyl-, hydrochloride (1:1)
InChI Formula InChI=1S/C19H25NO3.ClH/c1-14-11-19(23-4)15(12-18(14)22-3)9-10-20-13-16-7-5-6-8-17(16)21-2;/h5-8,11-12,20H,9-10,13H2,1-4H3;1H
SMILES Cl.COc1ccccc1CNCCc2cc(OC)c(C)cc2OC
IUPAC Name 2-(2,5-dimethoxy-4-methylphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine;hydrochloride
Molecular Weight (g/mol) 351.87
Formula Weight 351.1601
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.