missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxy-4-methylbenzaldehyde, 97%

Product Code. 15481209
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481209 25 g 25g
15461209 1 g 1g
15471209 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481209 Supplier Thermo Scientific Alfa Aesar Supplier No. H64140.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4925-88-6 | C10H12O3 | 180.20 g/mol

Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4925-88-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD02253192
InChI Key LRSRTWLEJBIAIT-UHFFFAOYSA-N
Synonym 4-methyl-2,5-dimethoxybenzaldehyde, unii-238b3l4auk, 2,5-dimethoxy-4-methyl-benzaldehyde, benzaldehyde, 2,5-dimethoxy-4-methyl, 2,5-dimethoxy-p-tolualdehyde, p-tolualdehyde, 2,5-dimethoxy, who-dd, 2,5-dimethoxy-4-methylbenzaldehyde;, 2,5-dimethoxy-4-methyl-benzaldehyde;, #
PubChem CID 602019
IUPAC Name 2,5-dimethoxy-4-methylbenzaldehyde
SMILES COC1=CC(C)=C(OC)C=C1C=O

Specifications

Melting Point 82°C to 86°C
Color White
Quantity 25 g
Beilstein 1866592
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 180.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dimethoxy-4-methylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.