missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimercapto-1,3,4-thiadiazole, Dipotassium Salt 98%, ACROS Organics™

Product Code. 10347960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347960 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347960 Supplier Thermo Scientific Acros Supplier No. 160820250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4628-94-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 270°C to 276°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C2K2N2S3
MDL Number MFCD00003104
Quantity 25 g
Beilstein 27, 677
Synonym 2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole
InChI Key GPWLFGDMYSVEGN-UHFFFAOYSA-L
SMILES C1(=NN=C(S1)[S-])[S-].[K+].[K+]
IUPAC Name dipotassium;1,3,4-thiadiazole-2,5-dithiolate
Molecular Weight (g/mol) 226.413
PubChem CID 78364
Formula Weight 226.41
Percent Purity ≥97.5%
Physical Form Crystalline Chunks or Powder
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole, dipotassium salt, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.