missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diiodothiophene, 99%

Product Code. 11469663
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469663 5 g 5g
11479663 25 g 25g
11489663 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469663 Supplier Thermo Scientific Alfa Aesar Supplier No. A12443.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-88-7 | C4H2I2S | 335.929 g/mol

2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diiodothiophene is used in the preparation of oligothiophene films and maskless fabrication of periodic patterns of a conjugated polymer. It finds application in the fabrication of oligothiophene and polythiophene micropatterns.

Solubility
Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-88-7
Molecular Formula C4H2I2S
Molecular Weight (g/mol) 335.929
MDL Number MFCD00014525
InChI Key PNYWRAHWEIOAGK-UHFFFAOYSA-N
Synonym thiophene, 2,5-diiodo, 2,5-diiodthiophen, thiophene,5-diiodo, pubchem5529, maybridge1_002904, acmc-1b0lo, 2,5-diiodothiophene, bidd:gt0169, thiophene, 2,5-diiodo-8ci 9ci
PubChem CID 69368
IUPAC Name 2,5-diiodothiophene
SMILES C1=C(SC(=C1)I)I

Specifications

Melting Point 39°C to 43°C
Boiling Point 139°C to 140°C (15 mmHg)
Quantity 5 g
Beilstein 109915
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 335.93
Percent Purity 99%
Chemical Name or Material 2,5-Diiodothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.