missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyterephthalic acid, 97%, Thermo Scientific Chemicals

Product Code. 15462289
Click to view available options
Quantity:
5 g
25 g

Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot dimethylformamide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-92-4
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00132933
InChI Key OYFRNYNHAZOYNF-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
PubChem CID 69131
IUPAC Name 2,5-dihydroxyterephthalic acid
SMILES C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O

Specifications

Melting Point >300°C
Color Gray to Green
Quantity 5 g
Beilstein 2696052
Solubility Information Soluble in hot dimethylformamide.
Formula Weight 198.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dihydroxyterephthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'