missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyphenylacetic acid, 97%, Thermo Scientific™

Product Code. 15437238
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437238 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15437238 Supplier Thermo Scientific Alfa Aesar Supplier No. H60742.ME
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an intermediate in the metabolism of tyrosine and phenylalanine.

Solubility
Soluble in methanol.

Notes
Store at cool temperature.
TRUSTED_SUSTAINABILITY

Specifications

CAS 451-13-8
Melting Point 150°C to 152°C
Molecular Formula C8H8O4
MDL Number MFCD00004324
Quantity 500 mg
Beilstein 2692860
Synonym homogentisic acid, 2,5-dihydroxyphenylacetic acid, alcapton, homogentisate acid, homogentisinic acid, 2,5-dihydroxyphenyl acetic acid, homogentisate, benzeneacetic acid, 2,5-dihydroxy, 2-2,5-dihydroxyphenyl acetic acid, 2,5-dihydroxy-alpha-toluic acid
Solubility Information Soluble in methanol.
InChI Key IGMNYECMUMZDDF-UHFFFAOYSA-N
SMILES OC(=O)CC1=CC(O)=CC=C1O
IUPAC Name 2-(2,5-dihydroxyphenyl)acetic acid
Molecular Weight (g/mol) 168.15
PubChem CID 780
ChEBI CHEBI:44747
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 2,5-Dihydroxyphenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.