missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid, TRC

Product Code. 30378337
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378337 Supplier Toronto Research Chemicals Supplier No. TRCD4516551G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gentisic Acid
CAS 490-79-9
Molecular Formula C7 H6 O4
Quantity 1 g
Synonym 2,5-Dihydroxybenzoic Acid, Mesalazine Imp. G (EP), 2,5-DHBA, 2,5-Dioxybenzoic acid, 3,6-Dihydroxybenzoic acid, 5-Hydroxysalicylic acid, Carboxyhydroquinone, Gensigen, Gensigon, Gentisic acid, Gentisinic acid, Hydroquinonecarboxylic acid, NSC 27224, NSC 49098, NSC 78825
InChI Formula InChI=1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)
SMILES OC(=O)c1cc(O)ccc1O
IUPAC Name 2,5-dihydroxybenzoic acid
Molecular Weight (g/mol) 154.12
Formula Weight 154.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.