missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid, Mikromol™

Product Code. 17942366
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942366 25 mg 25mg
14811351 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942366 Supplier Mikromol Supplier No. MM0217.100025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,5-Dihydroxybenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gentisic Acid
CAS 490-79-9
Analyte or Component Names Gentisic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O4
Certifications/Compliance ISO 17025
Synonym 2,5-Dihydroxybenzoic Acid, Mesalazine Imp. G (EP), 2,5-DHBA, 2,5-Dioxybenzoic acid, 3,6-Dihydroxybenzoic acid, 5-Hydroxysalicylic acid, Carboxyhydroquinone, Gensigen, Gensigon, Gentisic acid, Gentisinic acid, Hydroquinonecarboxylic acid, NSC 27224, NSC 49098, NSC 78825, Mesalazine Impurity G
InChI Formula InChI=1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)
SMILES OC(=O)c1cc(O)ccc1O
IUPAC Name 2,5-dihydroxybenzoic acid
Molecular Weight (g/mol) 154.12
Quantity 25 mg
Formula Weight 154.0266 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.