missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic acid, MedChemExpress

Product Code. 16499306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499306 Supplier Medchem Express Supplier No. HYW001179
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2,5-Dihydroxybenzoic acid is a derivative of benzoic and a powerful inhibitor of fibroblast growth factors.

2,5-Dihydroxybenzoic acid is a derivative of benzoic and a powerful inhibitor of fibroblast growth factors.

Specifications

Chemical Name or Material 2,5-Dihydroxybenzoic acid
CAS 490-79-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H6O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (648.85 mM)
SMILES OC(C=C1)=C(C(O)=O)C=C1O
Molecular Weight (g/mol) 154.12
Formula Weight 154.12
Percent Purity 99.93%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.