missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic acid, 99%

Product Code. 11490333
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490333 100 g 100g
11480333 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490333 Supplier Thermo Scientific Alfa Aesar Supplier No. A11459.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 490-79-9 | C7H6O4 | 154.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2,5-dihydroxybenzoic acid are used as intermediates for pharmaceuticals (especially for antipyetic anlgesic, antirheumatism) and other organic synthesis. They are used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field, they are used as intermediates for the production of other organic chemicals, resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals.

Solubility
It is soluble in alcohol and ether.

Notes
Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-79-9
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002460
InChI Key WXTMDXOMEHJXQO-UHFFFAOYSA-N
Synonym gentisic acid, hydroquinonecarboxylic acid, benzoic acid, 2,5-dihydroxy, 5-hydroxysalicylic acid, gensigen, gensigon, gentisate, 2,5-dioxybenzoic acid, 2,5-dhba, gentisinic acid
PubChem CID 3469
ChEBI CHEBI:17189
IUPAC Name 2,5-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)O

Specifications

Melting Point ∼206°C (decomposition)
Quantity 100 g
Beilstein 2209119
Merck Index 14,4398
Solubility Information It is soluble in alcohol and ether.
Formula Weight 154.12
Percent Purity 99%
Chemical Name or Material 2,5-Dihydroxybenzoic acid
compliance-icons
Product Identifier
  • 2,5-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,5-Dihydroxybenzoicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.