missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzaldehyde, 98+%

Product Code. 11413027
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413027 1 g 1g
11423027 5 g 5g
11433027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413027 Supplier Thermo Scientific Alfa Aesar Supplier No. A15565.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1194-98-5 | C7H6O3 | 138.12 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-98-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003333
InChI Key CLFRCXCBWIQVRN-UHFFFAOYSA-N
Synonym gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
PubChem CID 70949
ChEBI CHEBI:28508
IUPAC Name 2,5-dihydroxybenzaldehyde
SMILES OC1=CC=C(O)C(C=O)=C1

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Beilstein 1363961
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 138.12
Percent Purity ≥98%
Chemical Name or Material 2,5-Dihydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.