missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-1,4-benzoquinone, 98%

Product Code. 11464070
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464070 5 g 5g
11474070 25 g 25g
11430303 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464070 Supplier Thermo Scientific Alfa Aesar Supplier No. A11442.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-94-1 | C6H4O4 | 140.09 g/mol

2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-94-1
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00001598
InChI Key QFSYADJLNBHAKO-UHFFFAOYSA-N
Synonym 2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop
PubChem CID 69213
IUPAC Name 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC(=O)C(O)=CC1=O

Specifications

Melting Point ∼220°C (sublimation)
Quantity 5 g
Beilstein 1939229
Solubility Information Soluble in water.
Formula Weight 140.1
Percent Purity 98%
Chemical Name or Material 2,5-Dihydroxy-1,4-benzoquinone
compliance-icons
Product Identifier
  • 2,5-Dihydroxy-1,4-benzoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 2,5-Dihydroxy-1,4-benzoquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.