missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylacetic acid, 97%

Product Code. 11417214
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417214 5 g 5g
11407214 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417214 Supplier Thermo Scientific Alfa Aesar Supplier No. B21595.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 85068-27-5 | C8H6F2O2 | 172.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-27-5
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010000
InChI Key FKCRTRYQHZHXES-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetic acid, 2-2,5-difluorophenyl acetic acid, 2,5-difluorophenyl acetic acid, benzeneacetic acid, 2,5-difluoro, 2,5-difluorophenylaceticacid, pubchem4151, acmc-209xvw, ksc448m9d, 2,5-difluorophenyl-acetic acid, rarechem al bo 0248
PubChem CID 522823
IUPAC Name 2-(2,5-difluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1F)CC(=O)O)F

Specifications

Melting Point 123°C to 127°C
Quantity 5 g
Beilstein 6844956
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material 2,5-Difluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.