missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenol, 97%

Product Code. 11493018
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493018 5 g 5g
11403028 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493018 Supplier Thermo Scientific Alfa Aesar Supplier No. A18744.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2713-31-7 | C6H4F2O | 130.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2713-31-7
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00042501
InChI Key INXKVYFOWNAVMU-UHFFFAOYSA-N
Synonym phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464
PubChem CID 94952
IUPAC Name 2,5-difluorophenol
SMILES OC1=CC(F)=CC=C1F

Specifications

Melting Point 36°C to 40°C
Flash Point 53°C (127°F)
Odor Phenol-like
Quantity 5 g
UN Number UN2921
Beilstein 2081076
Formula Weight 130.09
Percent Purity 97%
Chemical Name or Material 2,5-Difluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.