missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzylamine, 97%, Thermo Scientific™

Product Code. 10615531 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10615531

Brand: Thermo Scientific Acros 311520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 85118-06-5
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.2200g/mL
Boiling Point 76°C to 78°C (15.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7F2N
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4940 to 1.496
MDL Number MFCD00010143
Quantity 1 g
Specific Gravity 1.22
Synonym 2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p
InChI Key GDFBHCMFIUBEQT-UHFFFAOYSA-N
SMILES NCC1=CC(F)=CC=C1F
IUPAC Name (2,5-difluorophenyl)methanamine
Molecular Weight (g/mol) 143.14
PubChem CID 123589
Formula Weight 143.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 5-Difluorobenzylamine, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.