missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzyl alcohol, 98%, Thermo Scientific™

Product Code. 11365289
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365289 5 g 5g
11375289 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365289 Supplier Thermo Scientific Alfa Aesar Supplier No. A19932.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75853-20-2
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010627
InChI Key KIKCAPPVFQLOIU-UHFFFAOYSA-N
Synonym 2,5-difluorobenzyl alcohol, 2,5-difluorophenyl methanol, benzenemethanol, 2,5-difluoro, 2,5-difluorobenzylalcohol, 2,5-difluorophenyl methan-1-ol, pubchem4913, acmc-209p0i, rarechem al bd 0214, 2,5-difluoro-phenyl-methanol
PubChem CID 522599
IUPAC Name (2,5-difluorophenyl)methanol
SMILES C1=CC(=C(C=C1F)CO)F

Specifications

Density 1.29
Flash Point 93°C (199°F)
Refractive Index 1.49
Quantity 5 g
Beilstein 9043529
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzyl alcohol
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.