missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoyl chloride, 97+%

Product Code. 11433394
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433394 5 g 5g
11443394 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433394 Supplier Thermo Scientific Alfa Aesar Supplier No. A10651.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35730-09-7 | C7H3ClF2O | 176.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35730-09-7
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00009929
InChI Key RLRUKKDFNWXXRT-UHFFFAOYSA-N
Synonym 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
PubChem CID 588082
IUPAC Name 2,5-difluorobenzoyl chloride
SMILES C1=CC(=C(C=C1F)C(=O)Cl)F

Specifications

Density 1.425
Boiling Point 81°C to 83°C (14 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.515
Quantity 5 g
UN Number UN3265
Beilstein 2046666
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.