missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzotrifluoride, 98%

Product Code. 15491159
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491159 1 g 1g
15401169 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491159 Supplier Thermo Scientific Alfa Aesar Supplier No. H64111.03
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 393-38-4 | C7H3F5 | 182.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-38-4
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.09
MDL Number MFCD00042494
InChI Key BLDOSVXRDMTZQE-UHFFFAOYSA-N
Synonym 2,5-difluorobenzotrifluoride, 1,4-difluoro-2-trifluoromethyl benzene, 1,4-difluoro-2-trifluoromethyl-benzene, benzene, 1,4-difluoro-2-trifluoromethyl, pubchem1650, 1-trifluoromethyl-2,5-difluorobenzene
PubChem CID 11111097
IUPAC Name 1,4-difluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(F)C=C1)C(F)(F)F

Specifications

Melting Point -14°C
Color Colorless to Yellow
Density 1.424 g/mL
Boiling Point 107°C to 108°C
Refractive Index 1.394
Quantity 1 g
Beilstein 2049494
Formula Weight 182.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.