missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzonitrile, 98+%

Product Code. p-7021611
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11427886

Brand: Thermo Scientific Alfa Aesar A14014.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64248-64-2 | C7H3F2N | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64248-64-2
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00001777
InChI Key OJTMHIMQUQOLJV-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-difluoro, 2,5-difluoro benzonitrile, 2,5-difluoro-benzonitrile, 2,5-difluorobenzenecarbonitrile, ncr bf ef, pubchem4754, 2,5-diflurobenzonitrile, 2,5 difluorobenzonitrile, 2,5-difluorobenzonitrile
PubChem CID 123558
IUPAC Name 2,5-difluorobenzonitrile
SMILES FC1=CC=C(F)C(=C1)C#N

Specifications

Melting Point 31°C to 35°C
Boiling Point 188°C
Flash Point 77°C (170°F)
Quantity 5 g
UN Number UN3439
Beilstein 2085640
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 2,5-Difluorobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.