missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 10175370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175370 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10175370 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 306750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2991-28-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 128°C to 132°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C7H4F2O2
Linear Formula F2C6H3COOH
MDL Number MFCD00002410
Quantity 1 g
Synonym benzoic acid, 2,5-difluoro, 2,5-difluorobenzoicacid, 2,5-difluoro-benzoic acid, pubchem4935, 2,5-difluorobenzic acid, acmc-1cj4a, ksc206g1b, rarechem al bo 0019, lbqmiavigllbgw-uhfffaoysa, 2,5-difluorobenzoic acid
InChI Key LBQMIAVIGLLBGW-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)C(=O)O)F
IUPAC Name 2,5-difluorobenzoic acid
Molecular Weight (g/mol) 158.104
PubChem CID 76339
Formula Weight 158.11
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Difluorobenzoic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.