missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoic acid, 98+%, Thermo Scientific™

Product Code. 11408753
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408753 25 g 25g
11488743 1 g 1g
11498743 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408753 Supplier Thermo Scientific Alfa Aesar Supplier No. A10503.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in the preparation of hydrazone derivatives. Acts as a potential antibacterial agent.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Difluorobenzoic acid is used in the preparation of hydrazone derivatives, which acts as a potential antibacterial agent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2991-28-8
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00002410
InChI Key LBQMIAVIGLLBGW-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-difluoro, 2,5-difluorobenzoicacid, 2,5-difluoro-benzoic acid, pubchem4935, 2,5-difluorobenzic acid, acmc-1cj4a, ksc206g1b, rarechem al bo 0019, lbqmiavigllbgw-uhfffaoysa, 2,5-difluorobenzoic acid
PubChem CID 76339
IUPAC Name 2,5-difluorobenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)F

Specifications

Melting Point 127°C to 132°C
Odor Odorless
Quantity 25 g
Beilstein 973351
Solubility Information Insoluble in water.
Formula Weight 158.11
Percent Purity ≥98%
Chemical Name or Material 2,5-Difluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.