missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzenesulfonamide, 98%, Thermo Scientific™

Catalog Number 15464147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
120022-63-1
C6H5F2NO2S
193.168
MFCD00798115
OLMFEUWXDKZGOO-UHFFFAOYSA-N
147822
2,5-difluorobenzenesulfonamide
C1=CC(=C(C=C1F)S(=O)(=O)N)F
This item is not returnable. View return policy
120022-63-1
C6H5F2NO2S
193.168
MFCD00798115
OLMFEUWXDKZGOO-UHFFFAOYSA-N
147822
2,5-difluorobenzenesulfonamide
C1=CC(=C(C=C1F)S(=O)(=O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 140°C to 142°C
Quantity 1 g
Synonym 2,5-difluorobenzenesulphonamide, 2,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 2,5-difluoro, 2,5-dfbs, acmc-20ai8p, 2,5-difluoro-benzenesulfonamide, benzenesulfonamide,2,5-difluoro, 2,5-bis fluoranyl benzenesulfonamide, 2,5-difluorobenzenesulfonamide
Formula Weight 193.17
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'