missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog Number 11414003
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
367-30-6
C6H5F2N
129.11
MFCD00007651
YNOOQIUSYGWMSS-UHFFFAOYSA-N
benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro
67775
2,5-difluoroaniline
C1=CC(=C(C=C1F)N)F
This item is not returnable. View return policy
367-30-6
C6H5F2N
129.11
MFCD00007651
YNOOQIUSYGWMSS-UHFFFAOYSA-N
benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro
67775
2,5-difluoroaniline
C1=CC(=C(C=C1F)N)F

2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 13°C to 14°C
Density 1.285
Boiling Point 171°C to 174°C
Flash Point 68°C (154°F)
Odor Characteristic
Refractive Index 1.513
Quantity 10 g
UN Number UN2941
Beilstein 2802549
Solubility Information Not miscible in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,5-Difluoroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'