missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoroaniline, 98%

Product Code. 11424003
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424003 50 g 50g
11414003 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424003 Supplier Thermo Scientific Alfa Aesar Supplier No. A10272.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-30-6 | C6H5F2N | 129.11 g/mol

2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-30-6
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007651
InChI Key YNOOQIUSYGWMSS-UHFFFAOYSA-N
Synonym benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro
PubChem CID 67775
IUPAC Name 2,5-difluoroaniline
SMILES C1=CC(=C(C=C1F)N)F

Specifications

Melting Point 13°C to 14°C
Density 1.285
Boiling Point 171°C to 174°C
Flash Point 68°C (154°F)
Odor Characteristic
Refractive Index 1.513
Quantity 50 g
UN Number UN2941
Beilstein 2802549
Solubility Information Not miscible in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,5-Difluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.